Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Hydroxypyrimidine-2-carbonitrile 97%

Supplier: AMBEED, INC
Warning

A597621-1G A597621-250MG A597621-5G
76819-372EA 145 USD InStock
76819-372 76819-374 76819-376
5-Hydroxypyrimidine-2-carbonitrile 97%
5-Hydroxypyrimidine-2-carbonitrile
Precaución: for Research Use Only


Formula: C₅H₃N₃O
MW: 121.09 g/mol
MDL Number: MFCD09608098
CAS Number: 345642-86-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty