Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Trifluoromethoxy)pyridine-2-carboxylic acid 95%

Supplier: AMBEED, INC
Danger

A600509-100MG A600509-250MG A600509-1G
76819-390EA 88 USD InStock
76819-390 76819-392 77673-614
5-(Trifluoromethoxy)pyridine-2-carboxylic acid 95%
5-(Trifluoromethoxy)pyridine-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₇H₄F₃NO₃
MDL Number: MFCD10697540
CAS Number: 102771-66-4
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty