Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ω,3,4-Trichloroacetophenone 97%

Supplier: AMBEED, INC
Danger

A612471-100G A612471-10G A612471-1G A612471-25G A612471-5G
76819-476EA 303 USD InStock
76819-476 76819-478 76819-480 76819-482 76819-484
ω,3,4-Trichloroacetophenone 97%
ω,3,4-Trichloroacetophenone
Precaución: for Research Use Only


Formula: C₈H₅Cl₃O
MW: 223.49
MDL Number: MFCD03970382
CAS Number: 42981-08-8
UN: 1759
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty