Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2,3-difluorobenzoic acid 95%

Supplier: AMBEED, INC
Warning

A618897-1G A618897-250MG A618897-5G
76819-502EA 60 USD InStock
76819-502 76819-504 77673-670
5-Bromo-2,3-difluorobenzoic acid 95%
5-Bromo-2,3-difluorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₃BrF₂O₂
MDL Number: MFCD04972472
CAS Number: 887585-64-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty