Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-6-(trifluoromethyl)benzoic acid 97%

Supplier: AMBEED, INC
Warning

A623939-100MG A623939-1G A623939-250MG
76819-540EA 14 USD InStock
76819-540 76819-542 76819-544
2-Methoxy-6-(trifluoromethyl)benzoic acid 97%
2-Methoxy-6-(trifluoromethyl)benzoic acid
Precaución: for Research Use Only


Formula: C₉H₇F₃O₃
MDL Number: MFCD09832319
CAS Number: 119692-41-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty