Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-6-(trifluoromethyl)pyridin-2-amine 96%

Supplier: AMBEED, INC
Warning

A625326-100MG A625326-1G A625326-250MG
76819-556EA 22 USD InStock
76819-556 76819-558 76819-560
3-Chloro-6-(trifluoromethyl)pyridin-2-amine 96%
3-Chloro-6-(trifluoromethyl)pyridin-2-amine
Precaución: for Research Use Only


Formula: C₆H₄ClF₃N₂
MW: 196.55 g/mol
MDL Number: MFCD04037237
CAS Number: 886762-09-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty