Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Fluoro-6-phenoxyphenyl)boronic acid 96%

Supplier: AMBEED, INC
Warning

A629799-100MG A629799-1G A629799-250MG
76819-594EA 14 USD InStock
76819-594 76819-596 76819-598
(2-Fluoro-6-phenoxyphenyl)boronic acid 96%
(2-Fluoro-6-phenoxyphenyl)boronic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₀BFO₃
MW: 232.01 g/mol
MDL Number: MFCD11856031
CAS Number: 1056372-58-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty