Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-β-Hydroxy-γ-butyrolactone 90%

Supplier: AMBEED, INC
Warning

A633606-10G A633606-1G A633606-250MG
76819-634EA 554 USD InStock
76819-634 76819-636 76819-638
(R)-(+)-β-Hydroxy-γ-butyrolactone 90%
(R)-(+)-β-Hydroxy-γ-butyrolactone
Precaución: for Research Use Only


Formula: C₄H₆O₃
MDL Number: MFCD00211248
CAS Number: 58081-05-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty