Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Pentenoic acid 98%

Supplier: AMBEED, INC
Danger

A649272-10G A649272-5G A649272-25G A649272-100G
76819-734EA 9 USD InStock
76819-734 76819-736 77308-060 77614-010
4-Pentenoic acid 98%
4-Pentenoic acid
Precaución: for Research Use Only


Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 187
Melting Pt: -18
MDL Number: MFCD00004408
CAS Number: 591-80-0
UN: 1760
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty