Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Diaminoterephthalic acid 95%

Supplier: AMBEED, INC
Warning

A657555-100MG A657555-1G A657555-250MG
76819-770EA 30 USD InStock
76819-770 76819-772 76819-774
2,5-Diaminoterephthalic acid 95%
2,5-Diaminoterephthalic acid
Precaución: for Research Use Only


Formula: C₈H₈N₂O₄
MDL Number: MFCD00464161
CAS Number: 945-30-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty