Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-chloro-1H-pyrrolo[2,3-b]pyridine-1-carboxylate 97%

Supplier: AMBEED, INC
Warning

A658617-100MG A658617-1G A658617-250MG
76819-780EA 18 USD InStock
76819-780 76819-782 76819-784
tert-Butyl 4-chloro-1H-pyrrolo[2,3-b]pyridine-1-carboxylate 97%
tert-Butyl 4-chloro-1H-pyrrolo[2,3-b]pyridine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₂H₁₃ClN₂O₂
MDL Number: MFCD11045066
CAS Number: 945599-50-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty