Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6,7-Dihydro-1H-pyrrolo[3,2-c]pyridin-4(5H)-one 95%

Supplier: AMBEED, INC
Warning

A665610-100MG A665610-1G A665610-250MG
76819-828EA 20 USD InStock
76819-828 76819-830 76819-832
6,7-Dihydro-1H-pyrrolo[3,2-c]pyridin-4(5H)-one 95%
6,7-Dihydro-1H-pyrrolo[3,2-c]pyridin-4(5H)-one
Precaución: For research use only.


Formula: C₇H₈N₂O
MDL Number: MFCD11506295
CAS Number: 736990-65-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty