Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Fluoro-3,4-dimethoxyphenyl)acetic acid 97%

Supplier: AMBEED, INC
Warning

A667083-250MG A667083-5G
76819-842EA 44 USD InStock
76819-842 76819-844
2-(2-Fluoro-3,4-dimethoxyphenyl)acetic acid 97%
2-(2-Fluoro-3,4-dimethoxyphenyl)acetic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₁FO₄
MW: 214.19 g/mol
MDL Number: MFCD03411454
CAS Number: 78495-65-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty