Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-tert-Butyl-3-hydroxycyclobutylcarbamate 97%

Supplier: AMBEED, INC
Warning

A673286-1G A673286-250MG A673286-5G
76819-882EA 12 USD InStock
76819-882 76819-884 76819-886
trans-tert-Butyl-3-hydroxycyclobutylcarbamate 97%
trans-tert-Butyl-3-hydroxycyclobutylcarbamate
Precaución: For research use only.


Formula: C₉H₁₇NO₃
MDL Number: MFCD09038209
CAS Number: 389890-42-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty