Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-5-methoxy-2-methylbenzenesulfonic acid 97%

Supplier: AMBEED, INC
Warning

A706209-100G A706209-25G A706209-500G A706209-5G
76820-064EA 24 USD InStock
76820-064 76820-066 76820-068 76820-070
4-Amino-5-methoxy-2-methylbenzenesulfonic acid 97%
4-Amino-5-methoxy-2-methylbenzenesulfonic acid
Precaución: for Research Use Only


Formula: C₈H₁₁NO₄S
MW: 217.24 g/mol
MDL Number: MFCD00035949
CAS Number: 6471-78-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty