Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Isobutyramidobenzeneboronic acid 98%

Supplier: AMBEED, INC
Warning

A731757-100MG A731757-1G A731757-250MG
76820-226EA 39 USD InStock
76820-226 76820-228 76820-230
4-Isobutyramidobenzeneboronic acid 98%
4-Isobutyramidobenzeneboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₄BNO₃
MDL Number: MFCD08235068
CAS Number: 874219-50-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty