Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-methylpyridine 98%

Supplier: AMBEED, INC
Danger

A748593-1000G A748593-100G A748593-25G A748593-500G
76820-302EA 74 USD InStock
76820-302 76820-304 76820-306 76820-308
2-Amino-4-methylpyridine 98%
2-Amino-4-methylpyridine
Precaución: For research use only.


Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 230.9
Melting Pt: 98
MDL Number: MFCD00006321
CAS Number: 695-34-1
UN: 2923
ADR: 8,6.1 Ⅱ
Merck Index: 13,00465
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty