Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-2-nitrobenzonitrile 96%

Supplier: AMBEED, INC
Danger

A760944-1G A760944-250MG A760944-25G A760944-5G
76820-344EA 19 USD InStock
76820-344 76820-346 76820-348 76820-350
3-Methyl-2-nitrobenzonitrile 96%
3-Methyl-2-nitrobenzonitrile
Precaución: for Research Use Only


Formula: C₈H₆N₂O₂
MW: 162.14 g/mol
MDL Number: MFCD09880086
CAS Number: 1885-77-4
UN: 3439
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty