Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dichloro(2-isopropoxyphenylmethylene)(tricyclohexylphosphine)ruthenium (II) 98%

Supplier: AMBEED, INC
Danger

A768276-1G A768276-250MG A768276-5G
76820-398EA 88 USD InStock
76820-398 76820-400 76820-402
Dichloro(2-isopropoxyphenylmethylene)(tricyclohexylphosphine)ruthenium (II) 98%
Dichloro(2-isopropoxyphenylmethylene)(tricyclohexylphosphine)ruthenium (II)
Precaución: for Research Use Only


Formula: C₂₈H₄₅Cl₂OPRu
MW: 600.60 g/mol
MDL Number: MFCD03453042
CAS Number: 203714-71-0
UN: 1325
ADR: 4.1 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty