Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Carboxy-5-chlorophenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A773168-100MG A773168-1G A773168-250MG A773168-5G
76820-416EA 10 USD InStock
76820-416 76820-418 76820-420 76820-422
3-Carboxy-5-chlorophenylboronic acid 98%
3-Carboxy-5-chlorophenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₆BClO₄
MW: 200.3841 g/mol
MDL Number: MFCD09258748
CAS Number: 957061-05-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty