Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-9H-purin-6-amine 98%

Supplier: AMBEED, INC
Warning

A794633-1G A794633-25G A794633-5G
76820-520EA 10 USD InStock
76820-520 76820-522 76820-524
2-Chloro-9H-purin-6-amine 98%
2-Chloro-9H-purin-6-amine
Precaución: for Research Use Only


Formula: C₅H₄ClN₅
MDL Number: MFCD00127773
CAS Number: 31458-56-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty