Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxy-5-methylphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A796511-100MG A796511-1G A796511-250MG
76820-536EA 37 USD InStock
76820-536 76820-538 76820-540
3-Methoxy-5-methylphenylboronic acid 98%
3-Methoxy-5-methylphenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₁₁BO₃
MW: 165.9821 g/mol
MDL Number: MFCD09952026
CAS Number: 725251-81-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty