Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Piperidino-5-pyrimidineboronic acid 98%

Supplier: AMBEED, INC
Warning

A847617-1G A847617-250MG A847617-5G
76820-732EA 90 USD InStock
76820-732 76820-734 76820-736
2-Piperidino-5-pyrimidineboronic acid 98%
2-Piperidino-5-pyrimidineboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₄BN₃O₂
MDL Number: MFCD07375149
CAS Number: 1002128-86-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty