Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

AZD-4547 98%

Supplier: AMBEED, INC
Warning

A868383-10MG A868383-1MG A868383-5MG A868383-100MG A868383-1G A868383-50MG
76820-800EA 22 USD InStock
76820-800 76820-802 76820-804 77674-292 77674-294 77674-296
AZD-4547 98%
AZD-4547
Precaución: for Research Use Only


Formula: C₂₆H₃₃N₅O₃
MW: 463.57 g/mol
MDL Number: MFCD22580423
CAS Number: 1035270-39-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty