Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tivantinib 98%

Supplier: AMBEED, INC
Danger

A891951-10MG A891951-1MG A891951-5MG
76820-896EA 32 USD InStock
76820-896 76820-898 76820-900
Tivantinib 98%
Tivantinib
Precaución: for Research Use Only


Formula: C₂₃H₁₉N₃O₂
MDL Number: MFCD11977597
CAS Number: 905854-02-6
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty