Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-(aminosulfonyl)thiophene-2-carboxylate 95%

Supplier: AMBEED, INC
Warning

A908249-100G A908249-1G A908249-250MG A908249-5G
76820-924EA 437 USD InStock
76820-924 76820-926 76820-928 76820-930
Methyl-3-(aminosulfonyl)thiophene-2-carboxylate 95%
Methyl-3-(aminosulfonyl)thiophene-2-carboxylate
Precaución: for Research Use Only


Formula: C₆H₇NO₄S₂
MDL Number: MFCD01760863
CAS Number: 59337-93-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty