Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Chlorophenoxy)acetic acid 98%

Supplier: AMBEED, INC
Warning

A909868-100G A909868-25G A909868-5G
76820-940EA 152 USD InStock
76820-940 76820-942 76820-944
(2-Chlorophenoxy)acetic acid 98%
(2-Chlorophenoxy)acetic acid
Precaución: for Research Use Only


Formula: (C₆H₄Cl)OCH₂COOH
MDL Number: MFCD00004298
CAS Number: 614-61-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty