Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(+)-Di-p-toluoyl-D-tartaric acid monohydrate 98%

Supplier: AMBEED, INC
Warning

A944246-1000G A944246-100G A944246-25G A944246-500G
76821-008EA 65 USD InStock
76821-008 76821-010 76821-012 76821-014
(+)-Di-p-toluoyl-D-tartaric acid monohydrate 98%
(+)-Di-p-toluoyl-D-tartaric acid monohydrate
Precaución: for Research Use Only


Formula: C₂₀H₁₈O₈·1H₂O
MDL Number: MFCD00149567
CAS Number: 71607-31-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty