Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-2-[4-(Trifluoromethyl)phenyl]vinylboronic acid 98%

Supplier: AMBEED, INC
Warning

A953566-100MG A953566-250MG
76821-042EA 35 USD InStock
76821-042 76821-044
trans-2-[4-(Trifluoromethyl)phenyl]vinylboronic acid 98%
trans-2-[4-(Trifluoromethyl)phenyl]vinylboronic acid
Precaución: for Research Use Only


Formula: C₉H₈BF₃O₂
MDL Number: MFCD03427005
CAS Number: 352525-91-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty