Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2,6-difluoropyridine 98%

Supplier: AMBEED, INC
Warning

A960252-100MG A960252-1G A960252-250MG
76821-066EA 49 USD InStock
76821-066 76821-068 76821-070
4-Bromo-2,6-difluoropyridine 98%
4-Bromo-2,6-difluoropyridine
Precaución: for Research Use Only


Formula: C₅H₂BrF₂N
MDL Number: MFCD21603855
CAS Number: 903513-58-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty