Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3,3,3-trifluoro-2,2-dimethylpropanoate 97%

Supplier: AMBEED, INC
Warning

A960727-100MG A960727-1G A960727-250MG A960727-5G
76821-072EA 5 USD InStock
76821-072 76821-074 76821-076 76821-078
Methyl 3,3,3-trifluoro-2,2-dimethylpropanoate 97%
Methyl 3,3,3-trifluoro-2,2-dimethylpropanoate
Precaución: for Research Use Only


Formula: C₆H₉F₃O₂
MDL Number: MFCD18253569
CAS Number: 1188911-72-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty