Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-D-His(Tos)-OH 98%

Supplier: AMBEED, INC
Warning

A979236-1G A979236-250MG A979236-5G
76821-112EA 43 USD InStock
76821-112 76821-114 76821-116
Boc-D-His(Tos)-OH 98%
Boc-D-His(Tos)-OH
Precaución: for Research Use Only


Formula: C₁₈H₂₃N₃O₆S
MDL Number: MFCD00037890
CAS Number: 69541-68-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty