Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromobenzamidine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A984799-1G A984799-25G A984799-5G
76821-134EA 22 USD InStock
76821-134 76821-136 76821-138
2-Bromobenzamidine hydrochloride 95%
2-Bromobenzamidine hydrochloride
Precaución: for Research Use Only


Formula: C₇H₈BrClN₂
MDL Number: MFCD04114438
CAS Number: 57075-82-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty