Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tozadenant 98+%

Supplier: AMBEED, INC
Warning

A100067-10MG A100067-1MG A100067-250MG A100067-50MG A100067-5MG
76821-182EA 46 USD InStock
76821-182 76821-184 76821-186 76821-188 76821-190
Tozadenant 98+%
Tozadenant
Precaución: for Research Use Only


Formula: C₁₉H₂₆N₄O₄S
MDL Number: MFCD21642026
CAS Number: 870070-55-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty