Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2,5-Dichloropyrimidin-4-yl)-1-(phenylsulfonyl)-1H-indole 97%

Supplier: AMBEED, INC
Warning

A103942-100MG A103942-250MG
76821-394EA 30 USD InStock
76821-394 76821-396
3-(2,5-Dichloropyrimidin-4-yl)-1-(phenylsulfonyl)-1H-indole 97%
3-(2,5-Dichloropyrimidin-4-yl)-1-(phenylsulfonyl)-1H-indole
Precaución: for Research Use Only


Formula: C₁₈H₁₁Cl₂N₃O₂S
MW: 404.27 g/mol
MDL Number: MFCD17392847
CAS Number: 882562-40-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty