Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-(bis(2,2,2-trifluoroethoxy)phosphoryl)acetate 97%

Supplier: AMBEED, INC
Warning

A104336-10G A104336-1G A104336-5G
76821-434EA 61 USD InStock
76821-434 76821-436 76821-438
Ethyl 2-(bis(2,2,2-trifluoroethoxy)phosphoryl)acetate 97%
Ethyl 2-(bis(2,2,2-trifluoroethoxy)phosphoryl)acetate
Precaución: for Research Use Only


Formula: C₈H₁₁F₆O₅P
MW: 332.13 g/mol
MDL Number: MFCD01318337
CAS Number: 124755-24-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty