Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Lauroyl-L-lysine 97%

Supplier: AMBEED, INC
Warning

A106569-10G A106569-1G A106569-5G
76821-578EA 712 USD InStock
76821-578 76821-580 76821-582 76821-584
N-Lauroyl-L-lysine 97%
N-Lauroyl-L-lysine
Precaución: for Research Use Only


Formula: C₁₈H₃₆N₂O₃
MW: 328.49 g/mol
MDL Number: MFCD00216730
CAS Number: 59409-41-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty