Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-(3-bromopropyl)benzoate 95%

Supplier: AMBEED, INC
Warning

A108724-100MG A108724-1G A108724-250MG A108724-5G
76821-678EA 19 USD InStock
76821-678 76821-680 76821-682 76821-684
Methyl 4-(3-bromopropyl)benzoate 95%
Methyl 4-(3-bromopropyl)benzoate
Precaución: for Research Use Only


Formula: C₁₁H₁₃BrO₂
MW: 257.12 g/mol
MDL Number: MFCD14708223
CAS Number: 113100-86-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty