Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GSK690693 98+%

Supplier: AMBEED, INC
Warning

A111141-10MG A111141-1MG A111141-5MG
76821-832EA 140 USD InStock
76821-832 76821-834 76821-836
GSK690693 98+%
GSK690693
Precaución: for Research Use Only


Formula: C₂₁H₂₇N₇O₃
MDL Number: MFCD14105605
CAS Number: 937174-76-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty