Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Morpholinopyridin-4-amine 98%

Supplier: AMBEED, INC
Warning

A112966-100MG A112966-250MG
76821-958EA 55 USD InStock
76821-958 76821-960
2-Morpholinopyridin-4-amine 98%
2-Morpholinopyridin-4-amine
Precaución: for Research Use Only


Formula: C₉H₁₃N₃O
MDL Number: MFCD02945666
CAS Number: 35980-77-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty