Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S,3S)-3-Amino-2-methyl-4-oxoazetidine-1-sulfonic acid 97%

Supplier: AMBEED, INC
Warning

A114393-100G A114393-1G A114393-25G A114393-500G A114393-5G
76822-028EA 103 USD InStock
76822-028 76822-030 76822-032 76822-034 76822-036
(2S,3S)-3-Amino-2-methyl-4-oxoazetidine-1-sulfonic acid 97%
(2S,3S)-3-Amino-2-methyl-4-oxoazetidine-1-sulfonic acid
Precaución: for Research Use Only


Formula: C₄H₈N₂O₄S
MDL Number: MFCD08061571
CAS Number: 80082-65-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty