Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 1-oxo-2,3-dihydro-1H-indene-5-carboxylate 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A117800-100MG A117800-250MG A117800-5G
76822-222EA 46 USD InStock
76822-222 76822-224 77683-059
Methyl 1-oxo-2,3-dihydro-1H-indene-5-carboxylate 95%, Ambeed.Inc
Methyl 1-oxo-2,3-dihydro-1H-indene-5-carboxylate
Precaución: For research use only.


Formula: C₁₁H₁₀O₃
MDL Number: MFCD09263429
CAS Number: 68634-02-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty