Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyano-2-iodobenzoic acid 95%

Supplier: AMBEED, INC
Warning

A118698-100MG A118698-250MG
76822-286EA 152 USD InStock
76822-286 76822-288
4-Cyano-2-iodobenzoic acid 95%
4-Cyano-2-iodobenzoic acid
Precaución: for Research Use Only


Formula: C₈H₄INO₂
MW: 273.027 g/mol
MDL Number: MFCD11847004
CAS Number: 1260814-20-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty