Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyano-2-phenylacetamide 98%

Supplier: AMBEED, INC
Warning

A119691-1G A119691-250MG A119691-5G
76822-330EA 89 USD InStock
76822-330 76822-332 76822-334
2-Cyano-2-phenylacetamide 98%
2-Cyano-2-phenylacetamide
Precaución: for Research Use Only


Formula: C₉H₈N₂O
MDL Number: MFCD17240132
CAS Number: 771-84-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty