Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-Cyanophenyl)guanidine 97%

Supplier: AMBEED, INC
Warning

A120413-10G A120413-1G A120413-25G
76822-370EA 46 USD InStock
76822-370 76822-372 76822-374
N-(4-Cyanophenyl)guanidine 97%
N-(4-Cyanophenyl)guanidine
Precaución: for Research Use Only


Formula: C₈H₈N₄
MDL Number: MFCD16251323
CAS Number: 5637-42-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty