Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

scriptaid 98%

Supplier: AMBEED, INC
Warning

A120568-10MG A120568-1MG A120568-50MG A120568-5MG
76822-386EA 29 USD InStock
76822-386 76822-388 76822-390 76822-392
scriptaid 98%
scriptaid
Precaución: for Research Use Only


Formula: C₁₈H₁₈N₂O₄
MDL Number: MFCD00386477
CAS Number: 287383-59-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty