Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-5-pyrimidineboronic acid 96%

Supplier: AMBEED, INC
Warning

A120798-100MG A120798-10G A120798-1G A120798-250MG A120798-5G
76822-400EA 9 USD InStock
76822-400 76822-402 76822-404 76822-406 76822-408
2-Methyl-5-pyrimidineboronic acid 96%
2-Methyl-5-pyrimidineboronic acid
Precaución: for Research Use Only


Formula: C₅H₇BN₂O₂
MW: 137.9323 g/mol
MDL Number: MFCD07375144
CAS Number: 1034924-06-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty