Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

TAE-226 98+%

Supplier: AMBEED, INC
Warning

A121000-10MG A121000-5MG
76822-418EA 197 USD InStock
76822-418 76822-420
TAE-226 98+%
TAE-226
Precaución: for Research Use Only


Formula: C₂₃H₂₅ClN₆O₃
MDL Number: MFCD12031516
CAS Number: 761437-28-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty