Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3-Ethylmorpholine hydrochloride 95%

Supplier: AMBEED, INC
Danger

A122255-100MG A122255-1G A122255-250MG A122255-5G
76822-500EA 22 USD InStock
76822-500 76822-502 76822-504 76822-506
(S)-3-Ethylmorpholine hydrochloride 95%
(S)-3-Ethylmorpholine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₄ClNO
MDL Number: MFCD11519097
CAS Number: 218594-88-8
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty