Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4,6-dihydropyrrolo[3,4-c]pyrazole-5(2H)-carboxylate 97%

Supplier: AMBEED, INC

A122289-10G A122289-1G A122289-250MG A122289-25G A122289-5G
76822-508EA 121 USD InStock
76822-508 76822-510 76822-512 76822-514 76822-516
tert-Butyl 4,6-dihydropyrrolo[3,4-c]pyrazole-5(2H)-carboxylate 97%
tert-Butyl 4,6-dihydropyrrolo[3,4-c]pyrazole-5(2H)-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₁₅N₃O₂
MDL Number: MFCD22565651
CAS Number: 1280210-79-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty